2'-C-Methyluridine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 1-((2R,3R,4R,5R)-3,4-Dihydroxy-5-(hydroxymethyl)-3-methyltetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-(2-C-Methyl-beta-D-ribofuranosyl)uracil

M3153-1G M3153-250MG
TCM3153-1GEA 78.28 USD
TCM3153-1G TCM3153-250MG
2'-C-Methyluridine ≥98.0% (by HPLC)
2'-C-Methyluridine
Formula: C₁₀H₁₄N₂O₆
MW: 258.23 g/mol
Melting Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02682960
CAS Number: 31448-54-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd5fe13b8654c214ddd49212f5e6ac46655cef14&vid=b495ea0b76dedc3f7daf508ecd9377ec1dd02e9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralisation titration) min. 95.0 %
Specific rotation [a]20/D +80.0 to +84.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR