6-Methoxy-4-chromanone ≥97.0% (by GC)

Supplier: TCI America

M3135-1G M3135-5G
TCM3135-1GEA 123.6 USD
TCM3135-1G TCM3135-5G
6-Methoxy-4-chromanone ≥97.0% (by GC)
6-Methoxy-4-chromanone
Formula: C₁₀H₁₀O₃
Boiling Pt: 180 °C
MDL Number: MFCD00190159
CAS Number: 5802-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc7d2e4c5e5c9afed32001e52b572deb3678a959&vid=45e2556b8a5a3ac1c1b9e939acf5baba51ea60a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR