Methyl-2-bromo-5-fluorobenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

M3101-25G M3101-5G
TCM3101-25GEA 290.46 USD
TCM3101-25G TCM3101-5G
Methyl-2-bromo-5-fluorobenzoate ≥98.0% (by GC)
Methyl-2-bromo-5-fluorobenzoate
Formula: C₈H₆BrFO₂
MDL Number: MFCD00086167
CAS Number: 6942-39-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58f20760d329d6d7edaa063298a056083ef782c5&vid=630ac95f3de49afd7014bd865d5eaaf40734f043&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 2.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR