Methyl p-tolyl sulfoxide ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1-Methyl-4-(methylsulphinyl)benzene, Methyl 4-Tolyl sulphoxide
M3061-1G
M3061-5G
TCM3061-1GEA
165.83
USD
TCM3061-1G
TCM3061-5G
Methyl p-tolyl sulfoxide ≥98.0% (by GC)
Methyl p-tolyl sulfoxide
Formula:
C₈H₁₀OS Boiling Pt: 114 °C |
MDL Number:
MFCD00075175 CAS Number: 934-72-5 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 40.0 to 44.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...