α,α,α-Trifluoro-p-xylene ≥98.0% (by GC)

Supplier: TCI America
Warning

M2945-25G M2945-5G
TCM2945-25GEA 276.04 USD
TCM2945-25G TCM2945-5G
α,α,α-Trifluoro-p-xylene ≥98.0% (by GC)
α,α,α-Trifluoro-p-xylene
Formula: C₈H₇F₃
Boiling Pt: 129 °C
Flash Pt: 27 °C
MDL Number: MFCD00075476
CAS Number: 6140-17-6
EINECS: 228-124-2
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42a00e556a25f1c35e5ecedb93d788737a65f3fe&vid=289086366ac98480a11e1f818205f67a4b12d3be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR