Dimethyl-3-thiaadipate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Methyl 3-[(2-Methoxy-2-oxoethyl)thio]propionate, Methyl 3-(Methoxycarbonylmethylthio)propionate, 3-[(2-Methoxy-2-oxoethyl)thio]propionic acid methyll ester, Dimethyl 3-Thiaadipate, 3-Thiaadipic acid dimethyl ester, 3-(Methoxycarbonylmethylthio)propionic acid methyll ester

M2922-1G M2922-5G
TCM2922-1GEA 38.11 USD
TCM2922-1G TCM2922-5G
Dimethyl-3-thiaadipate ≥98.0% (by GC)
Dimethyl-3-thiaadipate
Formula: C₇H₁₂O₄S
MW: 192.24 g/mol
Boiling Pt: 117 °C
Flash Pt: 54 °C
MDL Number: MFCD00043633
CAS Number: 7400-45-5
EINECS: 231-002-1
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11257fa23975f7cfd7ac91d4df513a49d32453a8&vid=6d6146de36e050b585b054aceb9fd9dd0e5181e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR