3-(4-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 5-(4-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde, 3-(4-Methoxyphenyl)pyrazole-4-carboxaldehyde

M2902-1G M2902-5G
TCM2902-1GEA 80.34 USD
TCM2902-1G TCM2902-5G
3-(4-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
3-(4-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde
Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
MDL Number: MFCD01133696
CAS Number: 199682-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dc10f06b6ad3255e20a6e4df73b61e534d1b45f&vid=c0c041e368f14e75c8719a76d9d3c77c8d08701e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 159.0 to 163.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR