methyl cyclobutanecarboxylate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Methyl cyclobutanecarboxylate

M2884-25G M2884-5G
TCM2884-25GEA 254.41 USD
TCM2884-25G TCM2884-5G
methyl cyclobutanecarboxylate ≥98.0% (by GC)
methyl cyclobutanecarboxylate
Formula: C₆H₁₀O₂
MW: 114.144 g/mol
Boiling Pt: 135 °C
MDL Number: MFCD00095142
CAS Number: 765-85-5
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df9c444d870c7eb7ad0be5fd27cb487a8b46fea7&vid=7ddceb97c611f4601522ccb979413860f0b458db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR