3-(Methylaminomethyl)benzonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

M2869-1G M2869-5G
TCM2869-1GEA 94.76 USD
TCM2869-1G TCM2869-5G
3-(Methylaminomethyl)benzonitrile ≥98.0% (by GC, titration analysis)
3-(Methylaminomethyl)benzonitrile
Formula: C₉H₁₀N₂
Boiling Pt: 145 °C
MDL Number: MFCD09731650
CAS Number: 90389-96-1
UN: 2922
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcca499f996977ff833896cc9364205c3b343609&vid=98939480d66cbdf9419696275965e6cc77b62977&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR