1-(4-Methoxyphenyl)cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-(4-Methoxyphenyl)-1-cyclopropanecarboxylic acid

M2837-25G M2837-5G
TCM2837-25GEA 547.96 USD
TCM2837-25G TCM2837-5G
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid
Formula: C₁₁H₁₂O₃
MW: 192.2112 g/mol
MDL Number: MFCD00019223
CAS Number: 16728-01-1
EINECS: 240-788-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralisation titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR