N-Methyl-N-[(trifluoromethyl)thio]-p-toluenesulfonamide ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

M2782-1G M2782-5G
TCM2782-1GEA 209.09 USD
TCM2782-1G TCM2782-5G
N-Methyl-N-[(trifluoromethyl)thio]-p-toluenesulfonamide ≥98.0% (by GC, total nitrogen)
N-Methyl-N-[(trifluoromethyl)thio]-p-toluenesulfonamide
Formula: C₉H₁₀F₃NO₂S₂
MW: 285.30 g/mol
MDL Number: MFCD29763020
CAS Number: 1045822-31-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ff8cbd66e83000dea2a7e52f618b734e2bfe9df&vid=f34a14047fd3df2bebbd2f39167381542dda68d6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR