2-Methylpropene ∼9% in hexane

Supplier: TCI America
Danger

I1083-100ML I1083-500ML
TCI1083-100MLEA 44.29 USD
TCI1083-100ML TCI1083-500ML
2-Methylpropene ∼9% in hexane
2-Methylpropene
Formula: C₄H₈
Storage Temperature: Refrigerator
MDL Number: MFCD00008898
CAS Number: 115-11-7
EINECS: 204-066-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 9.0 to 11.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR