4-Hydroxy-1-thiocoumarin ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Hydroxy-2H-1-benzothiopyran-2-one

H1597-1G H1597-5G
TCH1597-1GEA 115.36 USD
TCH1597-1G TCH1597-5G
4-Hydroxy-1-thiocoumarin ≥98.0% (by GC)
4-Hydroxy-1-thiocoumarin
Formula: C₉H₆O₂S
MW: 178.21 g/mol
MDL Number: MFCD00463527
CAS Number: 16854-67-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee32606603aa4adaedacdae3425d060e41e33588&vid=a7908f88436a24290d21db8ece37198216f002b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 213.0 to 217.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR