4-Fluoro-2-methoxy-5-nitroaniline ≥98.0% (by GC)

Supplier: TCI America
Warning

F1246-25G F1246-5G
TCF1246-25GEA 100.94 USD
TCF1246-25G TCF1246-5G
4-Fluoro-2-methoxy-5-nitroaniline ≥98.0% (by GC)
4-Fluoro-2-methoxy-5-nitroaniline
Formula: C₇H₇FN₂O₃
MW: 186.14 g/mol
MDL Number: MFCD23098941
CAS Number: 1075705-01-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a48b391b741272eb6c912fc27c94b17b0465b9b5&vid=2cf159018c4929bfe37adf378a150505124537e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 128.0 to 132.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR