Ethyl-5-methyl-1H-imidazole-4-carboxylate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Ethyl 4-Methyl-1H-imidazole-5-carboxylate, 4-Methyl-1H-imidazole-5-carboxylic acid ethyl ester, Ethyl 4-methyl-5-imidazolecarboxylate, Ethyl 5-methyl-1H-imidazole-4-carboxylate

E1386-25G E1386-5G
TCE1386-25GEA 170.98 USD
TCE1386-25G TCE1386-5G
Ethyl-5-methyl-1H-imidazole-4-carboxylate ≥98.0% (by HPLC)
Ethyl-5-methyl-1H-imidazole-4-carboxylate
Formula: C₇H₁₀N₂O₂
MW: 154.1665 g/mol
MDL Number: MFCD00005199
CAS Number: 51605-32-4
EINECS: 257-315-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0987cd395935f922394267b56d2bb18e509a7558&vid=43ce262162a00e1b27f88c6f3e3275305074e93b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 203.0 to 207.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR