α-Chloro-3,4-difluorotoulene ≥96.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 1-(Chloromethyl)-3,4-difluorobenzene, 3,4-Difluorobenzyl chloride, 4-(Chloromethyl)-1,2-difluorobenzene

D5117-25G D5117-5G
TCD5117-25GEA 196.73 USD
TCD5117-25G TCD5117-5G
α-Chloro-3,4-difluorotoulene ≥96.0% (by GC)
α-Chloro-3,4-difluorotoulene
Formula: C₇H₅ClF₂
MW: 162.57 g/mol
Boiling Pt: 80 °C
MDL Number: MFCD00236218
CAS Number: 698-80-6
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4915edf455d18d8311e12fb369f72673bb4e4c85&vid=aceff90f5e2c634f150b56e0a53963344d5f769e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR