6-Bromo-8-chloroimidazo[1,2-a]pyridine ≥98.0% (by GC)

Supplier: TCI America

B5848-1G B5848-5G
TCB5848-1GEA 90.64 USD
TCB5848-1G TCB5848-5G
6-Bromo-8-chloroimidazo[1,2-a]pyridine ≥98.0% (by GC)
6-Bromo-8-chloroimidazo[1,2-a]pyridine
Formula: C₇H₄BrClN₂
MDL Number: MFCD11518983
CAS Number: 474708-88-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f3a66561d9b7a54adf587cdf9bc141b6558b191&vid=4ad708b543ba17155aefb42a54c6a6e30e492859&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR