5-Bromo-2-fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 5-Bromo-2-fluoro-3-pyridylboronic, 5-Bromo-2-fluoro-3-pyridinylboronic acid, 5-Bromo-2-fluoropyridine-3-boronic acid

B5342-1G B5342-5G
TCB5342-1GEA 66.95 USD
TCB5342-1G TCB5342-5G
5-Bromo-2-fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)
5-Bromo-2-fluoro-3-pyridineboronic acid
Formula: C₅H₄BBrFNO₂
Melting Pt: 146 °C
MDL Number: MFCD04039317
CAS Number: 501435-91-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc51ef31054db57e0b6fc187a6f6a202a4532387&vid=fcc4c3f23a1a1e004b1ac125891791bbfd2e2801&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR