2-Butynyl toluene-4-sulfonate

Supplier: TCI America

Synonyms: 2-Butynyl p-toluenesulphonate, But-2-yn-1-yl 4-methylbenzenesulphonate

B4995-1G B4995-5G
TCB4995-1GEA 176.13 USD
TCB4995-1G TCB4995-5G
2-Butynyl toluene-4-sulfonate
2-Butynyl toluene-4-sulphonate
Formula: C₁₁H₁₂O₃S
MW: 224.276 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00078395
CAS Number: 56563-37-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR