5-Bromo-3-fluoropyridin-2-amine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A3138-1G A3138-200MG
TCA3138-1GEA 174.07 USD
TCA3138-1G TCA3138-200MG
5-Bromo-3-fluoropyridin-2-amine ≥98.0% (by GC, titration analysis)
5-Bromo-3-fluoropyridin-2-amine
Formula: C₅H₄BrFN₂
MDL Number: MFCD11848483
CAS Number: 748812-37-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e5b35ef3a048ccc88dde1c358cd58a2c7781ef2&vid=8dbba628482cc8e154c2014d4f4dace2e323f7c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 93.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR