Butyl acetate 99.5%, AcroSeal® for biochemistry
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate
447050010
447050025
TS447050010EA
107
USD
TS447050010
TS447050025
Butyl acetate 99.5%, AcroSeal® for biochemistry
Butyl acetate
Formula:
CH₃COO(CH₂)₃CH₃ MW: 116.16 g/mol Boiling Pt: 127 °C (760 mmHg) Melting Pt: –78 °C Density: 0.88 g/cm³ Flash Pt: 27 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009445 CAS Number: 123-86-4 UN: 1123 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.1 % |
GC | ≥99.4 % |
UV | at 400 nm A: ≤0.01 |
Color scale | ≤15 APHA |
Residue after evaporation | ≤0.01 % |
Heavy metals (as Pb) | ≤0.001 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...