Acetone 99.6% ACS

Supplier: Thermo Scientific Chemicals
Danger

400100010 400100025 400105000
TS400100010EA 96 USD
TS400100010 TS400100025 TS400105000
Acetone 99.6% ACS
Acetone
Formula: CH₃COCH₃
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.79 g/cm³
Flash Pt: –20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.5 % (K.F.)
Solubility (in water) Passes test
GC ≥99.5 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Isopropyl alcohol ≤0.05 %
Titrable acid ≤0.0003 meq/g
Titrable base ≤0.0006 meq/g
Aldehyde (HCHO) ≤0.002 %
Methanol ≤0.05 %
KMnO4-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR