Methyl ethyl ketone, extra dry 99.5%, AcroSeal®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Butanone
, MEK
, Butan-2-one
, EK 500 Additive
, Butanone
, Ethyl methyl ketone
396950010
396951000
TS396950010EA
438
USD
TS396950010
TS396951000
Methyl ethyl ketone, extra dry 99.5%, AcroSeal®
Methyl ethyl ketone
Formula:
C₂H₅COCH₃ MW: 72.11 g/mol Boiling Pt: 80 °C (760 mmHg) Melting Pt: –87 °C Density: 0.806 g/cm³ Flash Pt: –7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011648 CAS Number: 78-93-3 EINECS: 201-159-0 UN: 1193 ADR: 3,II REACH: 01-2119457290-43 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % |
GC | ≥99.4 % |
Residue after evaporation | ≤0.001 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...