Methyl ethyl ketone ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

389430010 389430025 389430100
TS389430010EA 76 USD
TS389430010 TS389430025 TS389430100
Methyl ethyl ketone ACS
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6
Melting Pt: -86
Density: 0.806 g/cm³
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3 II
REACH: 01-2119457290-43
Merck Index: 13,06097

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.20 %
GC ≥99.0 %
Color scale ≤15 APHA
Residue after evaporation ≤0.0025 %
Titrable acid ≤0.0005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR