Acetonitrile 99.9%, ECD tested Residual

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

326680010 326680025
TS326680010EA 83 USD
TS326680010 TS326680025
Acetonitrile 99.9%, ECD tested Residual
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Density: 0.781 g/cm³
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 % (Coulometric)
GC ≥99.9 %
Color scale ≤10 APHA
GC/ECD (as lindane) ≤5 ng/l
Residue after evaporation ≤0.0005 %
Acidity (CH3COOH) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR