1,2-Dichlorobenzene-[D₄] (98% D) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

321260010 321260050
TS321260010EA 65 USD
TS321260010 TS321260050
1,2-Dichlorobenzene-[D₄] (98% D) for NMR spectroscopy
1,2-Dichlorobenzene-[D₄]
Formula: C₆D₄Cl₂
Density: 1.34 g/cm³
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00037106
CAS Number: 2199-69-1
EINECS: 218-606-0
UN: 1591
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.2 % (Coulometric)
D-Enrichment ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR