iFluor® Ultra 750 succinimidyl ester
Supplier: AAT BIOQUEST INC
71680 71681 71682
77463-068EA 327.22 USD
77463-068 77463-070 77463-072
iFluor® Ultra 750 succinimidyl ester
iFluor® Ultra 750 succinimidyl ester
iFluor® Ultra 750 is a bright NIR fluorescent dye excitable by 633 nm to 685 nm laser lines, with a peak emission at 773 nm.
  • Stable for 12 months
Fluorescent dye-conjugated antibodies provide a tool for identifying proteins in many applications including fluorescent cell imaging, flow cytometry, western blotting, immunohistochemistry and more.
Precaución: For Research Use Only