iFluor® Ultra 647 succinimidyl ester
Supplier: AAT BIOQUEST INC
71671 71672 71670
77463-064EA 106.26 USD
77463-064 77463-066 77463-062
iFluor® Ultra 647 succinimidyl ester
iFluor® Ultra 647 succinimidyl ester
iFluor® Ultra 647 is an exceptionally bright far-red fluorescent dye excitable by 594 nm or 633 nm laser lines, with a peak emission at 670 nm.
  • Stable for 12 months
Fluorescent dye-conjugated antibodies provide a tool for identifying proteins in many applications including fluorescent cell imaging, flow cytometry, western blotting, immunohistochemistry and more.
Precaución: For Research Use Only