6-VIC phosphoramidite
Supplier: AAT Bioquest
6086 6087 6088
77462-968EA 258.75 USD
77462-968 77462-970 77462-972
6-VIC phosphoramidite
6-VIC phosphoramidite
6-VIC phosphoramidite is an asymmetrical xanthene dye widely used to prepare fluorescence-labeled oligonucleotides and design qPCR probes, such as TaqMan, Molecular Beacon, and Scorpion.
  • Stable for 6 months
Quality tested in-house
Precaución: For Research Use Only