Anti-IgG Goat Polyclonal Antibody (iFluor® 840)

Supplier: AAT BIOQUEST INC
48014 48015
77462-336EA 138.49 USD
77462-336 77462-338
Anti-IgG Goat Polyclonal Antibody (iFluor® 840)
Antibodies
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.

  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography

iFluor® 840 goat anti-mouse IgG (H+L) conjugates are ideal for various NIR imaging applications, including Western blotting, ELISA, protein arrays, tissue section imaging, and in vivo imaging.

Precaución: Research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 840
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR