2'-Deoxy-5-fluorocytidine >98%

Supplier: Sino Biological

Synonyms: 5-Fluoro-2'-deoxycytidine, 4-Amino-5-fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

D353-904D-100 D353-904D-20
77556-274EA 325 USD
77556-274 77556-276
2'-Deoxy-5-fluorocytidine >98%
2'-Deoxy-5-fluorocytidine
Formula: C₉H₁₂FN₃O₄
MW: 245.21 g/mol
Melting Pt: 196 °C
Storage Temperature: Freezer
MDL Number: MFCD00077348
CAS Number: 10356-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR