N-Phthalyl-L-tryptophan >98% (by TLC)

Supplier: Sino Biological

Synonyms: (S)-2-(1,3-Dioxoisoindolin-2-yl)-3-(1H-indol-3-yl)propanoic acid, RG 108, RG108

D353-904C-25 D353-904C-5
77556-270EA 275 USD
77556-270 77556-272
N-Phthalyl-L-tryptophan >98% (by TLC)
N-Phthalyl-L-tryptophan
Formula: C₁₉H₁₄N₂O₄
MW: 334.33 g/mol
Melting Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD08705332
CAS Number: 48208-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR