5-Aza-2'-deoxycytidine >98%

Supplier: Sino Biological

Synonyms: 5-Aza-2′-deoxycytidine, Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one, 2′-Deoxy-5-azacytidine 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one Decitabine, 4-Amino-1-(2-deoxy-beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one

D353-904B-10 D353-904B-50
77556-266EA 155 USD
77556-266 77556-268
5-Aza-2'-deoxycytidine >98%
5-Aza-2'-deoxycytidine
Formula: C₈H₁₂N₄O₄
MW: 228.21 g/mol
Melting Pt: 193...196 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043011
CAS Number: 2353-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR