GW2580 ≥98% (by TLC, NMR)

Supplier: Sino Biological

C74-901-05 C74-901-25
77555-804EA 155 USD
77555-804 77555-806
GW2580 ≥98% (by TLC, NMR)
GW2580
Formula: C₂₀H₂₂N₄O₃
Melting Pt: 209...210 °C
Storage Temperature: Ambient
MDL Number: MFCD12024701
CAS Number: 870483-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR