TG003 >98% (by HPLC, NMR)

Supplier: Sino Biological
Warning

C57-901-05 C57-901-25
77555-674EA 105 USD
77555-674 77555-676
TG003 >98% (by HPLC, NMR)
TG003
Formula: C₁₃H₁₅NO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00624584
CAS Number: 719277-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR