BML-259 >98% (by TLC, NMR)
Supplier: Sino Biological
|
|
C33-901-05
C33-901-25
77555-452EA
125
USD
77555-452
77555-454
BML-259 >98% (by TLC, NMR)
BML-259
Formula:
C₁₄H₁₆N₂OS Melting Pt: 150...152 °C Storage Temperature: Ambient |
MDL Number:
MFCD09802287 CAS Number: 267654-00-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...