KU55933 >98% (by TLC, NMR)

Supplier: Sino Biological

A26-901-05 A26-901-25
77554-360EA 83 USD
77554-360 77554-362
KU55933 >98% (by TLC, NMR)
KU55933
Formula: C₂₁H₁₇NO₃S₂
Storage Temperature: Freezer
MDL Number: MFCD08276985
CAS Number: 587871-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR