[-]-Deguelin >98% (by TLC, NMR)

Supplier: Sino Biological

A16-900B-05 A16-900B-25
77554-100EA 125 USD
77554-100 77554-102
[-]-Deguelin >98% (by TLC, NMR)
[-]-Deguelin
Formula: C₂₃H₂₂O₆
Storage Temperature: Ambient
MDL Number: MFCD01740600
CAS Number: 522-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR