PI(4,5)P2:PS Lipid Substrates
Supplier: Sino Biological
P429-59-1000 P429-59-500
77561-820EA 226 USD
77561-820 77561-822
PI(4,5)P2:PS Lipid Substrates
Assays Protein Assays
The substrate solution used for assaying lipid kinases.
Formulation: Frozen substrate solution containing 0.125 mM of Phosphatidylinositol 4,5-biphosphate [PI(4,5)P2 or PtdIns(4,5)P] and 1 mM of Phosphatidylserine (PS) in 1X lipid dilution buffer
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.