PI:PS Lipid Substrates
Supplier: Sino Biological
P425-59B-1000
P425-59B-500
77561-804EA
226
USD
77561-804
77561-806
PI:PS Lipid Substrates
Assays
Protein Assays
The substrate solution used for assaying lipid kinases.
Formulation: Frozen substrate solution containing 0.25 mM of Phospha-tidylinositol (PI) and 2 mM of Phosphatidylserine (PS) in 1X lipid dilution buffer
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.