PI(4)P Lipid Substrates
Supplier: Sino Biological
P422-59-1000 P422-59-500
77561-792EA 450 USD
77561-792 77561-794
PI(4)P Lipid Substrates
Assays Protein Assays
The substrate solution used for assaying lipid kinases.
Formulation: Frozen substrate solution containing 1 mM of Phospha-tidylinositol 4-phosphate [PI(4)P or PtdIns4P] in 1X lipid dilution buffer
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.