Magnetic Labels for BradyJet™ J7300 Color Label Printer
Supplier: Brady Worldwide
170450 170451
77581-108RL 105.46 USD
77581-108 77580-894
Magnetic Labels for BradyJet™ J7300 Color Label Printer
Labels
Experience effortless removal and repositioning with magnetic labels for the BradyJet™ J7300 Printer. Get the flexibility needed without adhesive residue or surface damage.
  • Non-adhesive
  • Matte finish
  • Magnetic label material
  • Service temperature range: –40 to +140 °F
Ideal for reusable, repositionable, and Lean/5S identification applications such as warehousing racks, bins, and storage. Magnetic labels facilitate visual workplace best practices, improve efficiency, and reduce errors in a wide variety of settings. Labels withstand moderate heat, mild chemicals, oil, and water. Easiest and quickest labeling option when removal and repositioning are frequent. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 colour Label Printer. Ink cartridges are required to print this material (use part numbers P-CMY-1 and P-K-1).