Paper Label Tapes for BradyJet™ J7300 Color Label Printer
Supplier: Brady Worldwide
176621 176622 176623 176587 176588 170471 170472
77581-052RL 147.75 USD
77581-052 77580-892 77580-900 77580-966 77581-030 77581-044 77580-940
Paper Label Tapes for BradyJet™ J7300 Color Label Printer
Labels
Low cost and high contrast, these inkjet receptive paper labels offer excellent smear resistance for economical light-duty identification.
  • Permanent acrylic adhesive
  • Matte finish
  • Service temperature range: −40 to 140 °F
Ideal for barcoding, quality and control (testing/production, circle labels, date/time), and warehousing racks, bins, and storage. Permanent acrylic adhesive ensures labels adhere strongly to a variety of flat and curved surfaces, including cardboard, plastic, and metal. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 Color Label Printer. Ink cartridges are required to print on this material (use part numbers P-CMY-1 and P-K-1).