Neddylation Assay Buffers
Supplier: Sino Biological
N01-09-01
N01-09-05
N01-09-20
77560-484EA
17
USD
77560-484
77560-486
77560-488
Neddylation Assay Buffers
Buffers
Assay Buffers
Stock solution of buffer used for assaying ubiquitin enzymes.
- Buffer passed through a 0.2 µm filter membrane and packaged aseptically
- Frozen solution of buffer containing: 50 mM HEPES, pH 7.5, 100 mM NaCl, 50 µg/ml BSA, 3 mM MgCl₂
- Store at −20 °C
For optimal storage, aliquot buffer into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...