Mini Industrial Vacuum Cleaners, VHS010-VHS011 ACD
Supplier: Nilfisk
4010100128 4010100129 4010100130
77574-838EA 3106.08 USD
77574-838 77574-834 77574-830
Mini Industrial Vacuum Cleaners, VHS010-VHS011 ACD
Vacuum Cleaners
The VHS010 and VHS011 ACD are a range of third-party ACD-certified vacuum cleaners, suitable for the collection of combustible dust and hazardous dust.
  • Lightweight and compact
  • Highly movable due to the 4 ball bearing pivoting wheels
  • Reliable with 900 W and a very good suction power
  • Metal-motorhead integrates the brushless bypass motor, safer and more robust in massive usage than standard moors, it has also an extended lifetime of up to 10.000 hours
  • Fully equipped with convenient handle, power cord holder, and accessories holder
  • Fully grounded and bounded,
More Product Information