Showa XC500 Cut-Resistant Industrial Gloves
Supplier: Showa
XC500L-08 XC500M-07 XC500S-06 XC500XL-09 XC500XXL-10
77575-148CS 448.56 USD
77575-148 77575-184 77575-174 77575-104 77575-176
Showa XC500 Cut-Resistant Industrial Gloves
Gloves
The ultra-light XC500 industrial glove combines a highly cut-resistant liner with a durable, abrasive-resistant polyurethane palm coating for use in various industrial applications.
  • Seamless HPPE/stainless steel/polyester liner
  • Durable, abrasive-resistant polyurethane palm coating
  • Seamless knit
  • Lightweight, form-fitting, and abrasion-resistant
Flexible and breathable XC500 industrial gloves provide increased dexterity in a form-fitting, seamless knit design. These gloves are recommended for use in maintenance, glass/window repair, component assembly, engineering, and mechanical industries.
Certificaciones: Meets ANSI level A5. EN 388:2016+A1:2018, EN ISO 21420:2020. CE marked.
Envase: Gloves are packaged 12 pair per polybag, 6 polybags per case (72 total pairs).