Brady® All-Weather Vinyl Labels
Supplier: Brady Worldwide
170833 170834 170835 170836 170837 170838 170839 170840 170841 170842 170823 170824 170825 170826 170827 170828 170829 170830 170831 170832 170943 170944 170945 170946 170947 170948 170949 170950 170843 170844 170845 170813 170814 170815 170816 170817 170818 170819 170820 170821 170822 170939 170940 170941 170942
76795-806EA 105.87 USD
76795-806 76795-952 76796-064 76795-860 76795-866 76796-012 76796-076 76796-188 76795-744 76795-956 76796-176 76795-712 76796-054 76795-754 76795-946 76795-848 76796-138 76796-026 76796-014 76796-020 76796-106 76795-936 76795-874 76796-024 76795-996 76796-086 76795-740 76796-074 76795-926 76795-896 76796-102 76796-174 76795-858 76796-218 76796-072 76795-750 76795-942 76796-042 76796-166 76795-902 76795-842 76795-794 76795-864 76795-992 76796-040
Brady® All-Weather Vinyl Labels
Labels
Versatile labels are ideal for labeling pipes, equipment, shelves, and other industrial surfaces.
  • Permanent acrylic adhesive
  • Gloss finish
  • Abrasion-resistant, chemical-resistant, oil-resistant, water-resistant material
  • Recommended ribbon: R6000HF, R4400
Tough labels are designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime. Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty, and smooth surfaces alike, especially in cold temperatures. Weather-resistant labels designed to last up to 10 years in an outdoor environment.
Certificaciones: BS 5609:1986 section 2 and section 3.