Brady® High Adhesion General Identification Polyester Labels
Supplier: Brady Worldwide
170772 170773 170771
76796-048EA 107.35 USD
76796-048 76795-786 76795-810
Brady® High Adhesion General Identification Polyester Labels
Labels
Versatile labels are perfect for general identification and marking powder-coated and smooth plastic surfaces.
  • Permanent rubber-based adhesive
  • Recommended ribbon: R6000HF
Durable labels are resistant to abrasions, extreme temperatures, and solvents such as fuel, oil, and water. Made with a permanent rubber-based adhesive that secures tightly to flat textured metals and smooth plastics alike.
Certificaciones: UL recognized to UL969, CSA accepted, RoHS compliant to 2011/65/EU (Pass 2015/863).