Brady® PermaSleeve® Heat-Shrink Labels
Supplier: Brady Worldwide
170712 170713 170714 170715 170716 170717 170718 170719 170720 170721 170722 170852 170853 170854 170855
76795-766EA 87.43 USD
76795-766 76795-870 76795-910 76796-066 76795-892 76796-126 76795-966 76796-080 76795-820 76795-876 76795-818 76795-822 76796-136 76796-002 76796-034
Brady® PermaSleeve® Heat-Shrink Labels
Labels
Specialized labels are designed specifically for rigorous cable and wire marking applications.
  • Matte finish
  • Flame-retardant, heat-shrinkable material
  • Recommended ribbon: R4300 (94000-380)
Durable labels are self-extinguishing and resistant to extreme temperatures, UV light, chemicals, fuel, and oil. Efficient and fast shrinking saves installation time and costs. Easily identifiable, these labels are made with a crisp, clear identification surface that’s resistant to abrasions and fading.
Certificaciones: UL recognised to UL224; SAE-AMS-DTL-23053/5 C (Class 1); SAE AS-81531.