Peptide Dilution Buffers
Supplier: Sino Biological
P58-09-01
P58-09-05
P58-09-20
77562-086EA
17
USD
77562-086
77562-088
77562-090
Peptide Dilution Buffers
Buffers
Assay Buffers
The working solution used for diluting peptide substrates.
Storage: For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...